2-Amino-3-bromopyridine

Warning

553719-25G 553719-5G
220077-983EA 224 USD
220077-983 220077-984
2-Amino-3-bromopyridine
2-Amino-3-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD03095187
CAS Number: 13534-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR