6-Bromo-3-methoxysalicylaldehyde ≥98%

Synonyms: 6-Bromo-2-hydroxy-3-methoxybenzaldehyde

553581-25G 553581-5G
220077-466EA 340.38 USD
220077-466 220077-467
6-Bromo-3-methoxysalicylaldehyde ≥98%
6-Bromo-3-methoxysalicylaldehyde
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00046152
CAS Number: 20035-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR