1-Butyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®

55292-1KG-F 55292-100G-F
220077-945EA 987.69 USD
220077-945 220077-944
1-Butyl-3-methylimidazoliumtetrachloroaluminate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium tetrachloroaluminate
Formula: C₈H₁₅AlCl₄N₂
MW: 308.01 g/mol
CAS Number: 80432-09-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR