3,5-Dimethylbenzenesulfonyl chloride ≥97%

Synonyms: 3,5-Dimethylbenzene-1-sulphonyl chloride, m-Xylene-5-sulphonyl chloride

552240-5G 552240-1G
220077-243EA 57.11 USD
220077-243 220077-242
3,5-Dimethylbenzenesulfonyl chloride ≥97%
3,5-Dimethylbenzenesulphonyl chloride
Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
MDL Number: MFCD03094655
CAS Number: 2905-27-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR