1-Butyl-3-methylimidazolium dicyanamide

Synonyms: 1-n-Butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyanamide

55220-5G-F 55220-50G-F
220077-826EA 146 USD
220077-826 220077-825
1-Butyl-3-methylimidazolium dicyanamide
1-Butyl-3-methylimidazolium dicyanamide
Formula: C₁₀H₁₅N₅
MW: 205.26 g/mol
Density: 1.06 g/cm³ (20 °C)
MDL Number: MFCD07784457
CAS Number: 448245-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR