5-Bromo-2-chloropyridine

Warning

551902-5G 551902-25G
220077-802EA 59.98 USD
220077-802 220077-801
5-Bromo-2-chloropyridine
5-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 71 °C
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR