5-Bromo-2,3-diaminopyridine

Synonyms: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

551899-5G 551899-25G
220077-800EA 59.55 USD
220077-800 220077-799
5-Bromo-2,3-diaminopyridine
5-Bromo-2,3-diaminopyridine
Formula: C₅H₆BrN₃
MW: 188.03 g/mol
Boiling Pt: 321 °C (1013 hPa)
MDL Number: MFCD00460094
CAS Number: 38875-53-5
EINECS: 254-172-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR