Vanadyl(IV) acetylacetonate

Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)

550787-10G 550787-50G
220077-734EA 74.77 USD
220077-734 220077-735
Vanadyl(IV) acetylacetonate
Vanadyl(IV) acetylacetonate
Formula: C₁₀H₁₄O₅V
MW: 265.16 g/mol
MDL Number: MFCD00000032
CAS Number: 3153-26-2
EINECS: 221-590-8
UN: 3285
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR