Silver methanesulfonate

550256-25G 550256-5G
220077-688EA 160.62 USD
220077-688 220077-689
Silver methanesulfonate
Silver methanesulphonate
Formula: CH₃AgO₃S
Melting Pt: 263 °C
MDL Number: MFCD00064795
CAS Number: 2386-52-9
EINECS: 219-199-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR