N,N,N',N'-Tetramethyl-1,3-propanediamine ≥99%

Danger

Synonyms: N,N,N',N'-Tetramethyl-1,3-diaminopropane, TMPDA, N1,N1,N3,N3-Tetramethylpropane-1,3-diamine, 1,3-Bis(dimethylamino)propane, N,N,N',N'-Tetramethyltrimethylenediamine

549983-2L 549983-100ML 549983-500ML
220077-659EA 314.46 USD
220077-659 220077-658 220077-660
N,N,N',N'-Tetramethyl-1,3-propanediamine ≥99%
1,3-Bis(dimethylamino)propane
Formula: C₇H₁₈N₂
MW: 130.23 g/mol
Boiling Pt: 145 °C (1013 hPa)
Density: 0.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008337
CAS Number: 110-95-2
EINECS: 203-818-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR