Tin(IV) oxide

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

549657-25G 549657-5G
220076-614EA 449.23 USD
220076-614 220076-615
Tin(IV) oxide
Tin(IV) oxide
Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1624.85 °C
Density: 6.95 g/cm³ (20 °C)
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR