3-Bromo-α,α,α,5-tetrafluorotoluene ≥97%

549029-5G 549029-1G
220077-557EA 340.62 USD
220077-557 220077-556
3-Bromo-α,α,α,5-tetrafluorotoluene ≥97%
3-Bromo-α,α,α,5-tetrafluorotoluene
Formula: C₇H₃BrF₄
MW: 243 g/mol
MDL Number: MFCD00042498
CAS Number: 130723-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR