2-Bromo-α,α,α,4-tetrafluorotoluene ≥97%

Synonyms: 2-Bromo-4-fluorobenzotrifluoride, 1-Bromo-3-fluoro-6-(trifluoromethyl)benzene

548995-5G 548995-1G
220077-552EA 340.62 USD
220077-552 220077-551
2-Bromo-α,α,α,4-tetrafluorotoluene ≥97%
2-Bromo-α,α,α,4-tetrafluorotoluene
Formula: C₇H₃BrF₄
MW: 243 g/mol
MDL Number: MFCD03094041
CAS Number: 351003-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR