5-Chloro-2-fluorobenzonitrile ≥97%

Warning

548693-1G 548693-5G
220077-531EA 26.92 USD
220077-531 220077-532
5-Chloro-2-fluorobenzonitrile ≥97%
5-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 120 °C (20 -rr)
Melting Pt: 67…70 °C (lit.)
Density: 1.584 g/cm³ (20 °C)
MDL Number: MFCD01631434
CAS Number: 57381-34-7
EINECS: 252-132-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR