2-Bromo-4-chloro-1-fluorobenzene ≥96%

Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene

548685-5G 548685-25G
220077-530EA 31.34 USD
220077-530 220077-529
2-Bromo-4-chloro-1-fluorobenzene ≥96%
2-Bromo-4-chloro-1-fluorobenzene
Formula: C₆H₃BrClF
MW: 209.44 g/mol
MDL Number: MFCD00672934
CAS Number: 1996-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR