2,4-Difluorobenzenesulfonyl chloride ≥97%

Synonyms: 2,4-Difluorobenzene-1-sulphonyl chloride

548081-5G 548081-25G
220077-442EA 51.08 USD
220077-442 220077-441
2,4-Difluorobenzenesulfonyl chloride ≥97%
2,4-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 1.581 g/cm³ (20 °C)
MDL Number: MFCD00042481
CAS Number: 13918-92-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR