3-Bromo-4-methylpyridine

Warning

Synonyms: 3-Bromo-4-picoline

548030-5G 548030-1G
220077-435EA 72.57 USD
220077-435 220077-434
3-Bromo-4-methylpyridine
3-Bromo-4-methylpyridine
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 47 - 48 °C (0.6 -rr)
Density: 1.549 g/cm³ (20 °C)
MDL Number: MFCD00082592
CAS Number: 3430-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR