3,5-Dichlorobenzenesulfonyl chloride ≥97%

Synonyms: 3,5-Dichlorobenzene-1-sulphonyl chloride

546933-5G 546933-25G
220077-303EA 43.85 USD
220077-303 220077-302
3,5-Dichlorobenzenesulfonyl chloride ≥97%
3,5-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
MW: 245.51 g/mol
Boiling Pt: 83 - 84 °C (0.1 -rr)
Melting Pt: 30 - 32 °C
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR