Butyl-4-hydroxybenzoate

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben

54680-1KG-F 54680-250G-F 54680-50G-F
220076-635EA 199.23 USD
220076-635 220076-636 220076-637
Butyl-4-hydroxybenzoate
Butyl-4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156 - 157 °C (3.5 -rr)
Melting Pt: 73 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
Merck Index: 13,01583

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR