Zinc bromide dihydrate

Danger

546739-100G 546739-500G
220075-781EA 90.46 USD
220075-781 220076-632
Zinc bromide dihydrate
Zinc bromide dihydrate
Formula: Br₂Zn·2H₂O
MW: 261.23 g/mol
Storage Temperature: Ambient
CAS Number: 18921-13-6
EINECS: 231-718-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR