Copper(II) phthalocyanine

Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

546682-2G 546682-200MG
220076-679EA 372.69 USD
220076-679 220076-678
Copper(II) phthalocyanine
Copper(II) phthalocyanine
Formula: C₃₂H₁₆CuN₈
MW: 576.08 g/mol
Melting Pt: 480 °C
MDL Number: MFCD00010719
CAS Number: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR