4-Amino-3,5-dichlorobenzoic acid ≥98%

Warning

545988-5G 545988-25G
220077-230EA 77.77 USD
220077-230 220077-229
4-Amino-3,5-dichlorobenzoic acid ≥98%
4-Amino-3,5-dichlorobenzoic acid
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
MDL Number: MFCD00017091
CAS Number: 56961-25-2
EINECS: 260-468-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR