3-Bromo-2-nitrotoluene ≥97%

Synonyms: 1-Bromo-3-methyl-2-nitrobenzene

545732-25G 545732-5G
220077-207EA 448.85 USD
220077-207 220077-208
3-Bromo-2-nitrotoluene ≥97%
3-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 25 - 29 °C
Density: 1.612 g/cm³ (25 °C)
MDL Number: MFCD02673631
CAS Number: 52414-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR