Ytterbium triacetate hydrate ≥99.999%

Synonyms: Ytterbium(III) acetate hydrate

544973-5G 544973-25G
220077-168EA 123.69 USD
220077-168 220077-167
Ytterbium triacetate hydrate ≥99.999%
Ytterbium triacetate hydrate
Formula: C₆H₉O₆Yb·H₂O
MW: 368.19 g/mol
MDL Number: MFCD00150127
CAS Number: 15280-58-7
EINECS: 244-137-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR