3-Fluoro-4-hydroxybenzoic acid ≥95%

544809-1G 544809-5G
220077-149EA 66.66 USD
220077-149 220077-150
3-Fluoro-4-hydroxybenzoic acid ≥95%
3-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00143161
CAS Number: 350-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR