Zirconium(IV) oxide

Warning

Synonyms: Zirconium dioxide, Zirconium Oxide

544760-25G 544760-5G
220077-146EA 235.38 USD
220077-146 220077-147
Zirconium(IV) oxide
Zirconium(IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: 2680 °C
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR