5-Chloro-2-thiophenesulfonyl chloride

Synonyms: 5-Chlorothiophene-2-sulphonyl chloride

544272-1G 544272-5G
220076-905EA 26.92 USD
220076-905 220076-906
5-Chloro-2-thiophenesulfonyl chloride
5-Chloro-2-thiophenesulphonyl chloride
Formula: C₄H₂Cl₂O₂S₂
MW: 217.1 g/mol
Boiling Pt: 64 - 66 °C (0.05 -rr)
Melting Pt: 28 °C
Density: 1.623 g/cm³ (20 °C)
MDL Number: MFCD00051667
CAS Number: 2766-74-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR