2,6-Difluorobenzenesulfonyl chloride ≥97%

Synonyms: 2,6-Difluorobenzene-1-sulphonyl chloride

544264-1G 544264-5G
220076-903EA 84 USD
220076-903 220076-904
2,6-Difluorobenzenesulfonyl chloride ≥97%
2,6-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 127.5 - 129.5 °C (1013 hPa)
Density: 1.568 g/cm³ (20 °C)
MDL Number: MFCD00085004
CAS Number: 60230-36-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR