5-Hexynoic acid ≥97%

Warning

544000-5G 544000-25G
220077-114EA 147.54 USD
220077-114 220077-113
5-Hexynoic acid ≥97%
5-Hexynoic acid
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 41 °C
Density: 1.0372 g/cm³ (20 °C)
MDL Number: MFCD00066346
CAS Number: 53293-00-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR