2-Chloroisonicotinic acid

Synonyms: 2-Chloro-4-pyridinecarboxylic acid, 2-Chloropyridine-4-carboxylic acid

543918-5G 543918-25G
220075-831EA 28.22 USD
220075-831 220075-830
2-Chloroisonicotinic acid
2-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR