2-Amino-3-bromo-5-methylpyridine

Warning

Synonyms: 3-Bromo-5-methylpyridin-2-amine, 2-Amino-3-bromo-5-picoline, 3-Bromo-5-methyl-2-pyridylamine

543071-1G 543071-5G
220077-100EA 35.43 USD
220077-100 220077-101
2-Amino-3-bromo-5-methylpyridine
2-Amino-3-bromo-5-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD00068231
CAS Number: 17282-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR