Lutetium trichloride hexahydrate ≥99.99%

542075-1G 542075-5G
220076-994EA 175.38 USD
220076-994 220076-995
Lutetium trichloride hexahydrate ≥99.99%
Lutetium trichloride hexahydrate
Formula: Cl₃Lu·6H₂O
MW: 389.42 g/mol
MDL Number: MFCD00011099
CAS Number: 15230-79-2
EINECS: 233-240-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR