2-Amino-4-chlorothiophenol ≥96%

Synonyms: 2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline

541850-5G 541850-25G
220076-987EA 55.85 USD
220076-987 220076-986
2-Amino-4-chlorothiophenol ≥96%
2-Amino-4-chlorothiophenol
Formula: C₆H₆ClNS
MW: 159.64 g/mol
MDL Number: MFCD00792528
CAS Number: 1004-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR