1,2-Dichloro-4-iodobenzene ≥98%

Synonyms: 3,4-Dichloroiodobenzene

541753-250G 541753-25G
220076-975EA 76.03 USD
220076-975 220076-976
1,2-Dichloro-4-iodobenzene ≥98%
1,2-Dichloro-4-iodobenzene
Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
MDL Number: MFCD00019014
CAS Number: 20555-91-3
EINECS: 243-874-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR