4,4,5,5,5-Pentafluoropentan-1-ol ≥95%

541133-5G 541133-25G
220076-877EA 66.02 USD
220076-877 220076-876
4,4,5,5,5-Pentafluoropentan-1-ol ≥95%
4,4,5,5,5-Pentafluoropentan-1-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
MDL Number: MFCD00153224
CAS Number: 148043-73-6
EINECS: 421-360-9

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR