2,4-Dichlorothiophenol ≥97%

Warning

Synonyms: 2,4-Dichlorobenzenethiol, 1,3-Dichloro-4-mercaptobenzene, 2,4-Dichlorobenzene-1-thiol

540862-5G 540862-1G
220076-848EA 88.46 USD
220076-848 220076-847
2,4-Dichlorothiophenol ≥97%
2,4-Dichlorothiophenol
Formula: C₆H₄Cl₂S
MW: 179.07 g/mol
Boiling Pt: 115 °C (8 mmHg)
Melting Pt: 22 °C
Density: 1.44 g/cm³ (20 °C)
MDL Number: MFCD00041421
CAS Number: 1122-41-4
EINECS: 214-346-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR