2'-Iodoacetophenone ≥97%

Synonyms: 1-(2-Iodophenyl)ethan-1-one, 1-(2-Iodophenyl)ethanone, o-Iodoacetophenone

540676-5G 540676-1G
220076-661EA 325.38 USD
220076-661 220076-660
2'-Iodoacetophenone ≥97%
2'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
MDL Number: MFCD00094998
CAS Number: 2142-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR