2,5-Dimethoxybenzylamine ≥97%

Synonyms: 1-(2,5-Dimethoxyphenyl)methanamine

540315-5G 540315-25G
220076-777EA 137.31 USD
220076-777 220076-776
2,5-Dimethoxybenzylamine ≥97%
2,5-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00052811
CAS Number: 3275-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR