(S)-(+)-1,2-Propanediol ≥96%

Synonyms: (S)-(+)-1,2-Propylene glykol, (S)-Propane-1,2-diol, (S)-(+)-1,2-Dihydroxy propane

540250-25G 540250-5G
220076-769EA 155.08 USD
220076-769 220076-770
(S)-(+)-1,2-Propanediol ≥96%
(S)-(+)-1,2-Propanediol
Formula: C₃H₈O₂
MW: 76.1 g/mol
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004539
CAS Number: 4254-15-3

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR