(R)-(-)-1,2-Propanediol ≥96%

Synonyms: (R)-(-)-1,2-Dihydroxy propane, (R)-(-)-1,2-Propylene glykol, (R)-Propane-1,2-diol

540242-5G 540242-25G
220076-767EA 37.69 USD
220076-767 220076-495
(R)-(-)-1,2-Propanediol ≥96%
(R)-(-)-1,2-Propanediol
Formula: C₃H₈O₂
MW: 76.1 g/mol
MDL Number: MFCD00066248
CAS Number: 4254-14-2

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR