(R)-(+)-1,2-Epoxypropane ≥99%

Synonyms: (R)-(+)-Propene oxide, (R)-(+)-Methyloxirane, (R)-(+)-1,2-Propylene oxide, (R)-(+)-Propylene oxide

540048-25G 540048-5G
220076-766EA 303.69 USD
220076-766 220076-468
(R)-(+)-1,2-Epoxypropane ≥99%
(R)-(+)-1,2-Epoxypropane
Formula: C₃H₆O
MW: 58.08 g/mol
Density: 0.829 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066211
CAS Number: 15448-47-2
UN: 1280
ADR: 3,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR