(S)-(-)-1,2-Epoxypropane ≥99%

Synonyms: (S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide

540021-25G 540021-5G
220076-763EA 359.69 USD
220076-763 220076-764
(S)-(-)-1,2-Epoxypropane ≥99%
(S)-(-)-1,2-Epoxypropane
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –112 °C
Density: 0.8268 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064312
CAS Number: 16088-62-3
EINECS: 240-241-0
UN: 1280
ADR: 3,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR