2-Fluoro-6-(trifluoromethyl)benzyl bromide ≥98%

Synonyms: 1-(Bromomethyl)-2-fluoro-6-(trifluoromethyl)benzene, 2-(Bromomethyl)-1-fluoro-3-(trifluoromethyl)benzene

539627-1G 539627-5G
220076-713EA 35.75 USD
220076-713 220076-714
2-Fluoro-6-(trifluoromethyl)benzyl bromide ≥98%
2-Fluoro-6-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 43 - 45 °C
MDL Number: MFCD00082477
CAS Number: 239087-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR