ω-Bromo-4-cyanoacetophenone ≥96%

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

539392-5G 539392-25G
220076-697EA 66.88 USD
220076-697 220076-696
ω-Bromo-4-cyanoacetophenone ≥96%
ω-Bromo-4-cyanoacetophenone
Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 92…94 °C
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR