3-Iodopyridine

Warning

539368-1G 539368-5G
220076-690EA 47.82 USD
220076-690 220076-691
3-Iodopyridine
3-Iodopyridine
Formula: C₅H₄IN
Boiling Pt: 90 - 92 °C (14 -rr)
Melting Pt: 53.5 °C
MDL Number: MFCD00023553
CAS Number: 1120-90-7
EINECS: 214-322-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR