α-Bromo-2-chloro-6-fluorotoluene ≥96%

Synonyms: 2-Chloro-6-fluorobenzyl bromide, 2-(Bromomethyl)-1-chloro-3-fluorobenzene

539090-1G 539090-5G
220075-784EA 194.85 USD
220075-784 220075-785
α-Bromo-2-chloro-6-fluorotoluene ≥96%
α-Bromo-2-chloro-6-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1.629 g/cm³ (20 °C)
MDL Number: MFCD00040126
CAS Number: 68220-26-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR