2-Amino-6-bromobenzothiazole

538507-1G 538507-5G 538507-25G
220076-560EA 30.8 USD
220076-560 220076-562 220076-561
2-Amino-6-bromobenzothiazole
2-Amino-6-bromobenzothiazole
Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR