cis-2-Hexene ≥95%

Synonyms: (Z)-2-Hexene

538493-1G 538493-5G
220076-558EA 68.71 USD
220076-558 220076-559
cis-2-Hexene ≥95%
cis-2-Hexene
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 67.8 °C (1013 hPa)
Melting Pt: –141.1 °C
Density: 0.6863 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066520
CAS Number: 7688-21-3
EINECS: 231-697-1
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR