3-Phenylbenzyl bromide ≥97%

Synonyms: 3-(Bromomethyl)biphenyl, m-(Bromomethyl)biphenyl

538388-25G 538388-5G
220076-147EA 426.46 USD
220076-147 220076-148
3-Phenylbenzyl bromide ≥97%
3-Phenylbenzyl bromide
Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
MDL Number: MFCD03452790
CAS Number: 14704-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR