2-Chloroethyl isocyanate ≥97%

Synonyms: Isocyanic acid 2-Chloroethyl ester

538337-5G 538337-25G
220076-140EA 32.31 USD
220076-140 220076-139
2-Chloroethyl isocyanate ≥97%
2-Chloroethyl isocyanate
Formula: C₃H₄ClNO
MW: 105.52 g/mol
Boiling Pt: 135 °C (1013 hPa)
Density: 1.24 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002043
CAS Number: 1943-83-5
EINECS: 217-734-4
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR